COMH4211AD4E Combi-Blocks, Inc.

3-(4-Fluoro-3-isopropoxy-phenyl)-2-methyl-propionic acid

  • MDL number MFCD31579317
  • Cas Number : 2270905-42-3
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2048
Availability details for
Quantity
  • Availability
  • Add to Cart