COMH4211AD4F Combi-Blocks, Inc.

tert-Butyl 3-(3-amino-2-methylphenyl)propanoate

  • MDL number MFCD31384971
  • Cas Number : 1191413-21-4
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2050
Availability details for
Quantity
  • Availability
  • Add to Cart