COMH4211AD50 Combi-Blocks, Inc.

5-(4-Chloro-2-fluorophenyl)pyrazin-2-amine

  • MDL number MFCD23852987
  • Cas Number : 1526887-70-6
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2051
Availability details for
Quantity
  • Availability
  • Add to Cart