COMH4211AD53 Combi-Blocks, Inc.

2-Bromo-3-phenylmethoxybenzaldehyde

  • MDL number MFCD30575616
  • Cas Number : 158584-99-7
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2054
Availability details for
Quantity
  • Availability
  • Add to Cart