COMH4211AD54 Combi-Blocks, Inc.

(R)-(1,2,3,4-Tetrahydro-naphthalen-1-yl)-urea

  • MDL number MFCD31613754
  • Cas Number : 2270918-29-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2055
Availability details for
Quantity
  • Availability
  • Add to Cart