COMH4211AD55 Combi-Blocks, Inc.

Methanesulfonic acid 2-tert-butoxycarbonylamino-2-methyl-propyl ester

  • MDL number MFCD20485977
  • Cas Number : 107017-68-5
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2057
Availability details for
Quantity
  • Availability
  • Add to Cart