COMH4211AD57 Combi-Blocks, Inc.

3-Fluoro-4-isobutoxybenzaldehyde

  • MDL number MFCD11156452
  • Cas Number : 1021231-45-7
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2059
Availability details for
Quantity
  • Availability
  • Add to Cart