COMH4211AD58 Combi-Blocks, Inc.

Chloromethyl 1-([(tert-butoxy)carbonyl]amino)cyclopropane-1-carboxylate

  • MDL number MFCD30732371
  • Cas Number : 1581704-70-2
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2061
Availability details for
Quantity
  • Availability
  • Add to Cart