COMH4211AD59 Combi-Blocks, Inc.

2-(4-Benzyloxy-pyrazol-1-yl)-phenylamine

  • MDL number MFCD31579545
  • Cas Number : 2270905-65-0
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2067
Availability details for
Quantity
  • Availability
  • Add to Cart