COMH4211AD5C Combi-Blocks, Inc.

3-(3,4-Dichlorophenoxy)propanamine

  • MDL number MFCD09883067
  • Cas Number : 164396-37-6
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2070
Availability details for
Quantity
  • Availability
  • Add to Cart