COMH4211AD5D Combi-Blocks, Inc.

4'-Fluoro-biphenyl-3-methanamine

  • MDL number MFCD05981710
  • Cas Number : 177976-50-0
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2073
Availability details for
Quantity
  • Availability
  • Add to Cart