COMH4211AD5F Combi-Blocks, Inc.

2-(4-((4'-Bromo-[1,1'-biphenyl]-4-yl)oxy)butoxy)tetrahydro-2h-pyran

  • MDL number MFCD31579500
  • Cas Number : 1620955-36-3
  • Molecular Weight : 405.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2076
Availability details for
Quantity
  • Availability
  • Add to Cart