COMH4211AD60 Combi-Blocks, Inc.

5-Chloro-2-(cyclopropylmethoxy)aniline

  • MDL number MFCD11136591
  • Cas Number : 1021105-81-6
  • Molecular Weight : 197.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2077
Availability details for
Quantity
  • Availability
  • Add to Cart