COMH4211AD61 Combi-Blocks, Inc.

6,6-Dimethyl-1,4,5,6-tetrahydrocyclopentapyrazole-3-carboxylic acid

  • MDL number MFCD23725892
  • Cas Number : 856256-47-8
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2079
Availability details for
Quantity
  • Availability
  • Add to Cart