COMH4211AD64 Combi-Blocks, Inc.

4-((1-Cyclobutylpiperidin-4-yl)oxy)aniline

  • MDL number MFCD27930300
  • Cas Number : 862310-33-6
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C15H22N2O/c16-12-4-6-14(7-5-12)18-15-8-10-17(11-9-15)13-2-1-3-13/h4-7,13,15H,1-3,8-11,16H2
Inchi Key CKZDBHBPTICGST-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2087
Availability details for
Quantity
  • Availability
  • Add to Cart