COMH4211AD66 Combi-Blocks, Inc.

4-[4-(2,5-Dioxo-pyrrolidin-1-yloxycarbonyl)-3-fluoro-phenyl]-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31579531
  • Cas Number : 2270911-87-8
  • Molecular Weight : 421.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2091
Availability details for
Quantity
  • Availability
  • Add to Cart