COMH4211AD68 Combi-Blocks, Inc.

Ethanol, 2-[2-[[(1,1-dimethylethyl)diphenylsilyl]oxy]ethoxy]-, 1-(4-methylbenzenesulfonate)

  • MDL number MFCD31561475
  • Cas Number : 1034710-40-1
  • Molecular Weight : 498.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-((tert-butyldiphenylsilyl)oxy)ethoxy)ethyl 4-methylbenzenesulfonate

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2095

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2095
Availability details for
Quantity
  • Availability
  • Add to Cart