COMH4211AD6A Combi-Blocks, Inc.

N-Benzyl-6-chloro-nicotinamide

  • MDL number MFCD03750288
  • Cas Number : 224817-12-3
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2098
Availability details for
Quantity
  • Availability
  • Add to Cart