COMH4211AD6B Combi-Blocks, Inc.

[4-Methyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridin-2-yl]-pyrrolidin-1-yl-methanone

  • MDL number MFCD31613898
  • Cas Number : 1951411-69-0
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2101
Availability details for
Quantity
  • Availability
  • Add to Cart