COMH4211AD6E Combi-Blocks, Inc.

2-Chloro-4-(2-fluoropyridin-4-yl)pyrimidine

  • MDL number MFCD08236719
  • Cas Number : 361147-25-3
  • Molecular Weight : 209.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2104
Availability details for
Quantity
  • Availability
  • Add to Cart