COMH4211AD6F Combi-Blocks, Inc.

2-Ethoxycarbonylmethoxy-5-iodo-benzoic acid methyl ester

  • MDL number MFCD31561593
  • Cas Number : 705262-57-3
  • Molecular Weight : 364.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2105
Availability details for
Quantity
  • Availability
  • Add to Cart