COMH4211AD71 Combi-Blocks, Inc.

(S)-1-Benzyl-4-glycidylpyrazole

  • MDL number MFCD31561610
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2108
Availability details for
Quantity
  • Availability
  • Add to Cart