COMH4211AD73 Combi-Blocks, Inc.

N-(3,4-Dichloro-benzyl)-o-methyl-hydroxylamine

  • MDL number MFCD21100670
  • Cas Number : 543730-44-5
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2114

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2114
Availability details for
Quantity
  • Availability
  • Add to Cart