COMH4211AD74 Combi-Blocks, Inc.

Trans propane-1-sulfonic acid (4-amino-cyclohexyl)-amide

  • MDL number MFCD31579394
  • Cas Number : 1867842-40-7
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2115
Availability details for
Quantity
  • Availability
  • Add to Cart