COMH4211AD75 Combi-Blocks, Inc.

2-(3-Chloro-benzyloxy)-5-fluoro-benzaldehyde

  • MDL number MFCD31557803
  • Cas Number : 1227622-75-4
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2117
Availability details for
Quantity
  • Availability
  • Add to Cart