COMH4211AD76 Combi-Blocks, Inc.

L-Valine, n-[(2-propen-1-yloxy)carbonyl]-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381659
  • Cas Number : 1343476-61-8
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2119
Availability details for
Quantity
  • Availability
  • Add to Cart