COMH4211AD77 Combi-Blocks, Inc.

2-Methyl-3-(3-trifluoromethoxy-phenyl)-propionic acid

  • MDL number MFCD23505377
  • Cas Number : 1542349-73-4
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2122
Availability details for
Quantity
  • Availability
  • Add to Cart