COMH4211AD78 Combi-Blocks, Inc.

1-Allyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583231
  • Cas Number : 2246635-45-8
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-allyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Inchi Code 1S/C14H23BN2O2/c1-8-9-17-11(3)12(10(2)16-17)15-18-13(4,5)14(6,7)19-15/h8H,1,9H2,2-7H3
Inchi Key YRMGNGSYBJRNMQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2126
Availability details for
Quantity
  • Availability
  • Add to Cart