COMH4211AD7A Combi-Blocks, Inc.

1-Bromo-3-(2,2-difluoroethoxy)-5-fluorobenzene

  • MDL number MFCD24083540
  • Cas Number : 1538752-02-1
  • Molecular Weight : 255.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2137
Availability details for
Quantity
  • Availability
  • Add to Cart