COMH4211AD7C Combi-Blocks, Inc.

2-(1-Allylpiperidin-4-yl)ethan-1-ol

  • MDL number MFCD29993925
  • Cas Number : 1863439-93-3
  • Molecular Weight : 169.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2139
Availability details for
Quantity
  • Availability
  • Add to Cart