COMH4211AD7D Combi-Blocks, Inc.

Cyclopropyl-(4-cyclopropylmethoxy-benzyl)-amine

  • MDL number MFCD11631124
  • Cas Number : 1095215-91-0
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2140
Availability details for
Quantity
  • Availability
  • Add to Cart