COMH4211AD7E Combi-Blocks, Inc.

1-(4-Bromo-2-nitrophenyl)-1h-imidazole

  • MDL number MFCD14652015
  • Cas Number : 23725-66-8
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2143
Availability details for
Quantity
  • Availability
  • Add to Cart