COMH4211AD80 Combi-Blocks, Inc.

3-Bromo-4-(2,2,2-trifluoroethoxy)pyridine

  • MDL number MFCD23162596
  • Cas Number : 1357095-12-5
  • Molecular Weight : 256.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2146
Availability details for
Quantity
  • Availability
  • Add to Cart