COMH4211AD81 Combi-Blocks, Inc.

N-Dihydrocinnamoylaminocaproic acid

  • MDL number MFCD00745034
  • Cas Number : 178622-38-3
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2147
Availability details for
Quantity
  • Availability
  • Add to Cart