COMH4211AD82 Combi-Blocks, Inc.

4-Amino-n-(3-benzyloxy-propyl)-butyramide

  • MDL number MFCD13320091
  • Cas Number : 1291526-70-9
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2152
Availability details for
Quantity
  • Availability
  • Add to Cart