COMH4211AD84 Combi-Blocks, Inc.

2-Ethoxy-4-(2-methoxy-ethoxy)-benzoic acid

  • MDL number MFCD31557874
  • Cas Number : 2204560-08-5
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2159
Availability details for
Quantity
  • Availability
  • Add to Cart