COMH4211AD85 Combi-Blocks, Inc.

(4-Ethoxy-3-fluorophenyl)methanamine

  • MDL number MFCD11191786
  • Cas Number : 1037164-60-5
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2160
Availability details for
Quantity
  • Availability
  • Add to Cart