COMH4211AD86 Combi-Blocks, Inc.

5-Bromo-2-(2,2,6,6-tetramethylpiperidin-4-yloxy)benzonitrile

  • MDL number MFCD31557901
  • Cas Number : 2058319-88-1
  • Molecular Weight : 337.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2163
Availability details for
Quantity
  • Availability
  • Add to Cart