COMH4211AD87 Combi-Blocks, Inc.

5-Bromo-2-cyclopropylmethoxy-n-methyl-benzamide

  • MDL number MFCD31628798
  • Cas Number : 2270909-26-5
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2164
Availability details for
Quantity
  • Availability
  • Add to Cart