COMH4211AD88 Combi-Blocks, Inc.

N-[(4-Bromo-2-chlorophenyl)methyl]cyclopropanamine

  • MDL number MFCD12065513
  • Cas Number : 1157720-31-4
  • Molecular Weight : 260.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2166
Availability details for
Quantity
  • Availability
  • Add to Cart