COMH4211AD89 Combi-Blocks, Inc.

(2-Amino-5-fluorophenyl)(pyrrolidin-1-yl)methanone

  • MDL number MFCD11132579
  • Cas Number : 1094290-32-0
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2168
Availability details for
Quantity
  • Availability
  • Add to Cart