COMH4211AD8A Combi-Blocks, Inc.

1-(2-Nitro-4-(trifluoromethyl)phenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD21337681
  • Molecular Weight : 383.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[2-nitro-4-(trifluoromethyl)phenyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Inchi Code 1S/C16H17BF3N3O4/c1-14(2)15(3,4)27-17(26-14)11-8-21-22(9-11)12-6-5-10(16(18,19)20)7-13(12)23(24)25/h5-9H,1-4H3
Inchi Key UCFTVLZEIMXNMZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2169
Availability details for
Quantity
  • Availability
  • Add to Cart