COMH4211AD8B Combi-Blocks, Inc.

2-(2-Benzyl-2h-tetrazol-5-yl)-phenol

  • MDL number MFCD31579362
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2171
Availability details for
Quantity
  • Availability
  • Add to Cart