COMH4211AD8C Combi-Blocks, Inc.

2-Methyl-3-(3-methylphenyl)propan-1-ol

  • MDL number MFCD20386003
  • Cas Number : 119052-84-5
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2173

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2173
Availability details for
Quantity
  • Availability
  • Add to Cart