COMH4211AD8D Combi-Blocks, Inc.

Propanoic acid, 3-[(diphenylmethyl)thio]-

  • MDL number MFCD31381707
  • Cas Number : 63547-34-2
  • Molecular Weight : 272.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2175
Availability details for
Quantity
  • Availability
  • Add to Cart