COMH4211AD8F Combi-Blocks, Inc.

5-(4,4-Difluoropiperidin-1-yl)picolinaldehyde

  • MDL number MFCD28013652
  • Cas Number : 1707365-72-7
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2177
Availability details for
Quantity
  • Availability
  • Add to Cart