COMH4211AD90 Combi-Blocks, Inc.

2,2-Difluoro-2-(p-tolyl)ethanol

  • MDL number MFCD30184094
  • Cas Number : 1783961-43-2
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2178

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2178
Availability details for
Quantity
  • Availability
  • Add to Cart