COMH4211AD91 Combi-Blocks, Inc.

2-Bromo-5-(2-bromoethoxy)pyridine

  • MDL number MFCD22987892
  • Cas Number : 1660157-37-8
  • Molecular Weight : 280.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2181
Availability details for
Quantity
  • Availability
  • Add to Cart