COMH4211AD92 Combi-Blocks, Inc.

1-Amino-n-(cyclopropylmethyl)cyclobutane-1-carboxamide hydrochloride

  • MDL number MFCD31539548
  • Cas Number : 2206607-55-6
  • Molecular Weight : 204.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2182
Availability details for
Quantity
  • Availability
  • Add to Cart