COMH4211AD93 Combi-Blocks, Inc.

2-(Ethylsulfanyl)-5-methylaniline

  • MDL number MFCD11645644
  • Cas Number : 65523-85-5
  • Molecular Weight : 167.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2184
Availability details for
Quantity
  • Availability
  • Add to Cart